753807
CdSeS/ZnS alloyed quantum dots
fluorescence λem 665 nm, 6 nm diameter, 1 mg/mL in toluene
Synonym(s):
QD alkyl functionalized, Fluorescent Nanocrystals, Trilite™
About This Item
Recommended Products
form
liquid
concentration
1 mg/mL in toluene
refractive index
n20/D 1.496 (lit.)
diameter
6 nm
bp
110-111 °C (lit.)
density
0.865 g/mL at 25 °C (lit.)
fluorescence
λem 665 nm
functional group
oleic acid
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Analysis Note
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
39.2 °F
Flash Point(C)
4 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.
Professor Sharma and colleagues review the synthesis and applications of this novel material. This includes a discussion of the unique properties of quantum dots and their suitability for solar cell applications, along with common synthesis techniques used to develop these materials.
Professor Xiaohu Gao (University of Washington, USA) provides a overview of recent quantum dot (QD) advancements and their potential for advancing bioassay and bioimaging technologies.
Perovskite quantum dots research progresses overcoming challenges, enabling rapid development of light-emitting devices.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service