Skip to Content
Merck
All Photos(1)

Key Documents

752312

Sigma-Aldrich

2′-Hydroxyspiro[norbornene-2,5′- [1,3,2]-dioxaphosphorinane]-2′-oxide

96%

Synonym(s):

5-Norbornene-2,2-dimethanol cyclic phosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13O4P
Molecular Weight:
216.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

188-193 °C

functional group

phosphate

SMILES string

OP1(=O)OCC2(CO1)C[C@H]3C[C@@H]2C=C3

InChI

1S/C9H13O4P/c10-14(11)12-5-9(6-13-14)4-7-1-2-8(9)3-7/h1-2,7-8H,3-6H2,(H,10,11)/t7-,8+/m1/s1

InChI key

KFMPMTRRWRSWAY-SFYZADRCSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service