Skip to Content
Merck
All Photos(1)

Documents

739715

Sigma-Aldrich

Glycochenodeoxycholic-2,2,4,4-d4 acid

98 atom % D, 98% (CP)

Synonym(s):

3α,7α-Dihydroxy-5β-cholanoic-2,2,4,4-d4 acid N-(carboxymethyl)amide, N-(3α,7α-Dihydroxy-24-oxocholan-24-yl)glycine-2,2,4,4-d4, Chenodeoxycholylglycine-2,2,4,4-d4, Chenodeoxyglycocholic-2,2,4,4-d4 acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26D4H39NO5
CAS Number:
Molecular Weight:
453.65
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

liquid

mass shift

M+4

SMILES string

[H][C@@]12[C@]([C@](CC([C@H]3O)([2H])[2H])(C)[C@@](C[C@H]2O)([H])C3([2H])[2H])([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCC(NCC(O)=O)=O

InChI

1S/C25H41NO6/c1-24-9-7-15(27)11-14(24)12-20(29)23-17-4-3-16(25(17,2)10-8-18(23)24)19(28)5-6-21(30)26-13-22(31)32/h14-20,23,27-29H,3-13H2,1-2H3,(H,26,30)(H,31,32)/t14-,15+,16+,17-,18-,19+,20+,23-,24-,25+/m0/s1/i7D2,11D2

InChI key

XILGPPBPXPUXQG-LXAYGWTLSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Le Zhan et al.
American journal of physiology. Gastrointestinal and liver physiology, 310(2), G93-G102 (2015-11-14)
Long-term parenteral nutrition (PN) administration can lead to PN-associated liver diseases (PNALD). Although multiple risk factors have been identified for PNALD, to date, the roles of bile acids (BAs) and the pathways involved in BA homeostasis in the development and
Diana M Shih et al.
Journal of lipid research, 56(1), 22-37 (2014-11-08)
We performed silencing and overexpression studies of flavin containing monooxygenase (FMO) 3 in hyperlipidemic mouse models to examine its effects on trimethylamine N-oxide (TMAO) levels and atherosclerosis. Knockdown of hepatic FMO3 in LDL receptor knockout mice using an antisense oligonucleotide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service