Skip to Content
Merck
All Photos(1)

Documents

727466

Sigma-Aldrich

4-Azidotoluene solution

~0.5 M in tert-butyl methyl ether, ≥95.0% (HPLC)

Synonym(s):

1-Azido-4-methylbenzene solution, 4-Methylphenyl azide solution, p-Tolyl azide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N3
CAS Number:
Molecular Weight:
133.15
Beilstein:
1841411
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (HPLC)

form

liquid

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

−20°C

SMILES string

Cc1ccc(cc1)N=[N+]=[N-]

InChI

1S/C7H7N3/c1-6-2-4-7(5-3-6)9-10-8/h2-5H,1H3

InChI key

YVXDRCDGBCOJHX-UHFFFAOYSA-N

General description

4-Azidotoluene is an aromatic azide generally used in copper(I)-catalyzed azide-alkyne cycloaddition reactions.

Application

4-Azidotoluene solution may be used for the functionalization at the axial position of boron subphthalocyanines with a terminal acetylene functionality via copper(I)-catalyzed azide-alkyne cycloaddition in the presence of Hunig′s base.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-18.4 °F

Flash Point(C)

-28 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Acetylenic Scaffolding with Subphthalocyanines.
Gotfredsen H, et al.
European Journal of Organic Chemistry, 2016(1), 17-21 (2016)
Ju Eun Jeon et al.
Cell, 180(1), 176-187 (2020-01-11)
In response to biotic stress, plants produce suites of highly modified fatty acids that bear unusual chemical functionalities. Despite their chemical complexity and proposed roles in pathogen defense, little is known about the biosynthesis of decorated fatty acids in plants.
Cu-catalyzed azide? alkyne cycloaddition
Meldal M & Torn?e CW
Chemical Reviews, 108(8), 2952-3015 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service