Skip to Content
Merck
All Photos(1)

Key Documents

700029

Sigma-Aldrich

SIMes-leucinol

97%

Synonym(s):

1-(S)-Leucinol-3-(2,4,6-trimethylphenyl)-3H-imidazolium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H27F6N2OP
Molecular Weight:
432.38
UNSPSC Code:
12352101
PubChem Substance ID:

Assay

97%

form

powder

reaction suitability

reaction type: click chemistry
reagent type: catalyst

mp

168-170 °C

storage temp.

2-8°C

SMILES string

F[P-](F)(F)(F)(F)F.CC(C)C[C@@H](CO)[N+]1=CN(CC1)c2c(C)cc(C)cc2C

InChI

1S/C18H29N2O.F6P/c1-13(2)8-17(11-21)19-6-7-20(12-19)18-15(4)9-14(3)10-16(18)5;1-7(2,3,4,5)6/h9-10,12-13,17,21H,6-8,11H2,1-5H3;/q+1;-1/t17-;/m0./s1

InChI key

NTQWYGMQOMHOQG-LMOVPXPDSA-N

Application

Chiral NHC used with copper for conjugate addition of Grignards to enones

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Martin et al.
Journal of the American Chemical Society, 128(26), 8416-8417 (2006-06-29)
The copper-catalyzed asymmetric conjugate addition of Grignard reagents to trisubstituted cyclic enones affords enantioenriched all-carbon quaternary centers with up to 96% ee. The chiral ligand is a diaminocarbene, directly generated in situ. The combination of Grignard reagent and diaminocarbene is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service