Skip to Content
Merck
All Photos(1)

Key Documents

694657

Sigma-Aldrich

Lumidot CdSe/ZnS

510, core-shell type quantum dots, 5 mg/mL in toluene

Synonym(s):

QD, Quantum dot semiconductor nanocrystal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
To inquire about this product 694657, please contact your local Merck office or dealer. Contact Technical Service

form

dispersion

availability

not available in USA

concentration

5 mg/mL in toluene

particle size

3.0 nm

bp

110 °C

density

0.865 g/mL at 25 °C

ε (extinction coefficient)

0.045 x 105 M-1cm-1

fluorescence

λex 495 nm; λem 510 nm FWHM <40 nm, quantum yield ≥30%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Pale green emitting quantum dot nanoparticles

Legal Information

Product of Nanoco Technologies sold under US and worldwide patents.
Lumidot is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3

Flash Point(F)

closed cup

Flash Point(C)

closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service