Skip to Content
Merck
All Photos(1)

Key Documents

688002

Sigma-Aldrich

Poly[(2,5-didecyloxy-1,4-phenylene) (2,4,6-triisopropylphenylborane)], diphenyl terminated

Synonym(s):

Boramer-TC03, Boramer-T03, Poly(2,5-didecyloxy-1,4-phenylene tripylborane), diphenyl terminated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103

form

solid

mol wt

average Mn ~1446

fluorescence

λex 300 nm; λem 486 nm in chloroform

Application

n-type polymer semiconductor; HOMO = –6.2 eV, LUMO = –3.0 eV.

Legal Information

Manufactured by TDA research, Inc.
Boramer is a trademark of TDA Research, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashwani Kumar Gupta et al.
Journal of biological methods, 8(2), e150-e150 (2021-07-15)
The prevalence of kidney dysfunction continues to increase worldwide, driving the need to develop transplantable renal tissues. The kidney develops from four major renal progenitor populations: nephron epithelial, ureteric epithelial, interstitial and endothelial progenitors. Methods have been developed to generate
Human iPSC-derived hepatocytes in 2D and 3D suspension culture for cryopreservation and in vitro toxicity studies.
Altmaier, et al.
Reproductive Toxicology, 111, 68-80 (2022)
Hendrik Schürmann et al.
Journal of cell science, 135(10) (2022-05-28)
Podosomes are mechanosensitive protrusive actin structures that are prominent in myeloid cells, and they have been linked to vascular extravasation. Recent studies have suggested that podosomes are hierarchically organized and have coordinated dynamics on the cell scale, which implies that
Luebben, S.; Sapp, S.
Material Matters, 3, 11-11 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service