Skip to Content
Merck
All Photos(1)

Documents

686239

Sigma-Aldrich

Zirconium acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zr(CH2CHCO2)4
CAS Number:
Molecular Weight:
375.44
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

reaction suitability

reagent type: catalyst
core: zirconium

SMILES string

C=CC(=O)O[Zr](OC(=O)C=C)(OC(=O)C=C)OC(=O)C=C

InChI

1S/4C3H4O2.Zr/c4*1-2-3(4)5;/h4*2H,1H2,(H,4,5);/q;;;;+4/p-4

InChI key

CXNQUHPJUJGOHC-UHFFFAOYSA-J

Application

Please see product no. 686204 for additional application and usage information (Aldrich Technical Bulletin AL-238 )
Zirconium-containing multifunctional acrylate useful for producing cured, transparent films with high refractive indices.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service