Skip to Content
Merck
All Photos(1)

Documents

674109

Sigma-Aldrich

(S)-(+)-3-Aminopiperidine dihydrochloride

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2 · 2HCl
CAS Number:
Molecular Weight:
173.08
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

96%

form

solid

mp

205-209 °C

SMILES string

Cl.Cl.N[C@H]1CCCNC1

InChI

1S/C5H12N2.2ClH/c6-5-2-1-3-7-4-5;;/h5,7H,1-4,6H2;2*1H/t5-;;/m0../s1

InChI key

GGPNYXIOFZLNKW-XRIGFGBMSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service