639273
Methanol-13C,d2
97 atom % 13C, 98 atom % D, 98% (CP)
Synonym(s):
Methanol-13C, d2 (Methyl alcohol), Methyl-13C,d2 alcohol
About This Item
Recommended Products
isotopic purity
97 atom % 13C
98 atom % D
Assay
98% (CP)
bp
65.4 °C (lit.)
density
0.865 g/mL at 25 °C
mass shift
M+3
SMILES string
[2H][13CH]([2H])O
InChI
1S/CH4O/c1-2/h2H,1H3/i1+1D2
InChI key
OKKJLVBELUTLKV-WGVGGRBOSA-N
Related Categories
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
52.0 °F - closed cup
Flash Point(C)
11.1 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service