Skip to Content
Merck
All Photos(2)

Key Documents

634069

Sigma-Aldrich

N-Boc-2-bromobenzylamine

97%

Synonym(s):

N-(tert-Butoxycarbonyl)-2-bromobenzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH2NHCO2C(CH3)3
CAS Number:
Molecular Weight:
286.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

50-54 °C (lit.)

functional group

amine
bromo

SMILES string

CC(C)(C)OC(=O)NCc1ccccc1Br

InChI

1S/C12H16BrNO2/c1-12(2,3)16-11(15)14-8-9-6-4-5-7-10(9)13/h4-7H,8H2,1-3H3,(H,14,15)

InChI key

DFNZFCPEUDSNEO-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service