Skip to Content
Merck
All Photos(1)

Key Documents

606790

Sigma-Aldrich

Benzene-12C6

99.9 atom % 12C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
12C6H6
Molecular Weight:
78.05
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

description

13C-depleted

isotopic purity

99.9 atom % 12C

Assay

99% (CP)

bp

80 °C (lit.)

mp

5.5 °C (lit.)

density

0.873 g/mL at 25 °C

mass shift

depleted

SMILES string

[12cH]1[12cH][12cH][12cH][12cH][12cH]1

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H/i1+0,2+0,3+0,4+0,5+0,6+0

InChI key

UHOVQNZJYSORNB-WDBXEELJSA-N

General description

Benzene-12C6 is also known as depleted benzene.

Application

Benzene-12C6 may be employed in FTIR (Fourier transform infrared spectroscopy) measurements.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F

Flash Point(C)

-11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John C Thomas et al.
ACS nano, 9(5), 4734-4742 (2015-04-14)
Carboranethiol molecules self-assemble into upright molecular monolayers on Au{111} with aligned dipoles in two dimensions. The positions and offsets of each molecule's geometric apex and local dipole moment are measured and correlated with sub-Ångström precision. Juxtaposing simultaneously acquired images, we
Xavier Moreel et al.
Cancer prevention research (Philadelphia, Pa.), 7(7), 766-776 (2014-05-16)
The association between omega-3 (ω-3) fatty acids and prostate cancer has been widely studied. However, little is known about the impact of prostate tissue fatty acid content on prostate cancer progression. We hypothesized that compared with the estimated dietary ω-3
H Muchalski et al.
Organic & biomolecular chemistry, 13(4), 1249-1253 (2014-12-02)
The peroxidation of 7-dehydrocholesterol (7-DHC), a biosynthetic precursor to vitamin D3 and cholesterol, has been linked to the pathophysiology of Smith-Lemli-Optiz syndrome (SLOS), a devastating human disorder. In SLOS, 7-DHC plasma and tissue levels are elevated because of defects in
Paula Mayorga Burrezo et al.
Journal of the American Chemical Society, 137(11), 3834-3843 (2015-03-03)
We have used Raman spectroscopy to study the molecular and electronic structures of the radical cations and dications of carbon-bridged oligo(para-phenylenevinylene)s (COPVn, n = 1-6) possessing consecutive fused pentagons and hexagons, up to 19, along with COPV derivatives having electron-donating
Emma Oakton et al.
Dalton transactions (Cambridge, England : 2003), 43(40), 15138-15142 (2014-09-03)
Herein, we report the preparation of small and narrowly distributed (2.1 ± 0.5 nm) Ag nanoparticles supported on passivated silica, where the surface OH groups are replaced by OSiMe3 functionalities. This synthetic method involves the grafting of silver(I) bis(trimethylsilyl)amide ([AgN(SiMe3)2]4)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service