Skip to Content
Merck
All Photos(1)

Key Documents

575704

Sigma-Aldrich

Tetraamminepalladium(II) tetrachloropalladate(II)

99.999%

Synonym(s):

NSC 212525

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Pd(NH3)4](PdCl4)
CAS Number:
Molecular Weight:
422.77
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.999%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

192 °C (dec.) (lit.)

density

2.489 g/mL at 25 °C (lit.)

SMILES string

N.N.N.N.[Pd++].Cl[Pd--](Cl)(Cl)Cl

InChI

1S/4ClH.4H3N.2Pd/h4*1H;4*1H3;;/q;;;;;;;;2*+2/p-4

InChI key

SAZHJAFUUQUXRW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Studies involving
  • The thermal decomposition of ammonium and ammine chloro complexes of palladium
  • Bactericidal activities
  • Anticaner activities

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service