Skip to Content
Merck
All Photos(1)

Documents

566101

Sigma-Aldrich

Bis(indenyl)halfnium(IV) dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C9H7)2HfCl2
CAS Number:
Molecular Weight:
479.70
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

reaction suitability

core: hafnium
reagent type: catalyst

mp

211-215 °C (dec.) (lit.)

SMILES string

Cl[Hf]Cl.[CH]1[CH][C]2C=CC=C[C]2[CH]1.[CH]3[CH][C]4C=CC=C[C]4[CH]3

InChI

1S/2C9H7.2ClH.Hf/c2*1-2-5-9-7-3-6-8(9)4-1;;;/h2*1-7H;2*1H;/q;;;;+2/p-2

InChI key

YVAJTQPCQNUSIG-UHFFFAOYSA-L

Application

  • Cocatalyst in dehydrogenation of cyclooctane

Catalyst for:
  • CH activation reactions
  • Metalation of Bronsted acids
  • Chain termination reactions in ethene polymerization

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service