563412
1-(1-Naphthyl)-2-(trimethylsilyl)acetylene
97%
Synonym(s):
Trimethyl[(naphthalen-1-yl)ethynyl]silane
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
C10H7C≡CSi(CH3)3
CAS Number:
Molecular Weight:
224.37
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
refractive index
n20/D 1.595 (lit.)
bp
292-293 °C (lit.)
density
0.968 g/mL at 25 °C (lit.)
SMILES string
C[Si](C)(C)C#Cc1cccc2ccccc12
InChI
1S/C15H16Si/c1-16(2,3)12-11-14-9-6-8-13-7-4-5-10-15(13)14/h4-10H,1-3H3
InChI key
WATBCTJRCLNXNF-UHFFFAOYSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service