474886
2-Chloro-5-(trifluoromethyl)phenyl isothiocyanate
98%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
ClC6H3(CF3)NCS
CAS Number:
Molecular Weight:
237.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
refractive index
n20/D 1.57 (lit.)
bp
239 °C (lit.)
density
1.447 g/mL at 25 °C (lit.)
SMILES string
FC(F)(F)c1ccc(Cl)c(c1)N=C=S
InChI
1S/C8H3ClF3NS/c9-6-2-1-5(8(10,11)12)3-7(6)13-4-14/h1-3H
InChI key
KHTMKXDMVYHDSY-UHFFFAOYSA-N
General description
2-Chloro-5-(trifluoromethyl)phenyl isothiocyanate is an organic building block. Its enthalpy of vaporization at boiling point (512.15K) has been reported to be 45.053kjoule/mol.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 548-548 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service