Skip to Content
Merck
All Photos(1)

Documents

463248

Sigma-Aldrich

Glycolic acid ethoxylate lauryl ether

average Mn ~460

Synonym(s):

Laureth-6 carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11-13(OCH2CH2)nOCH2CO2H
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mn ~460

impurities

≤1 wt. % NaCl

refractive index

n20/D 1.452

density

1 g/mL at 25 °C (lit.)

SMILES string

OCCO.OCC(O)=O.CCCCCCCCCCCCO.CCCCCCCCCCCCCO.CCCCCCCCCCCCCCO

InChI

1S/C14H30O.C13H28O.C12H26O.C2H4O3.C2H6O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15;1-2-3-4-5-6-7-8-9-10-11-12-13-14;1-2-3-4-5-6-7-8-9-10-11-12-13;3-1-2(4)5;3-1-2-4/h15H,2-14H2,1H3;14H,2-13H2,1H3;13H,2-12H2,1H3;3H,1H2,(H,4,5);3-4H,1-2H2

InChI key

KQSLVJFTAGAPJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Anionic surfactant.
Shampoos and encapsulated bath products.

Features and Benefits

Mild, non-irritating surfactant.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service