Skip to Content
Merck
All Photos(1)

Documents

44089

Sigma-Aldrich

Boc-Lys(2-Cl-Z)-PAM resin

extent of labeling: ~0.6 mmol/g loading

Synonym(s):

Nα-Boc-Nε-(2-chloro-Z)-L-lysine bound to PAM resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

extent of labeling

~0.6 mmol/g loading

particle size

100-200 mesh

storage temp.

2-8°C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service