Skip to Content
Merck
All Photos(1)

Documents

433209

Sigma-Aldrich

Sodium chloride

≥99%, AR grade

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

AR grade

Assay

≥99%

form

solid

availability

available only in EU

mp

801 °C (lit.)

solubility

water: soluble 358 g/L at 20 °C

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium chloride (NaCl) may be used in the preparation of alkaline Congo Red solution and as a reagent for the estimation of iodine in water samples.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eagleson M.
Concise Encyclopedia Chemistry, 996-996 (1994)
Suppression of methane/air explosion by ultrafine water mist containing sodium chloride additive.
Cao X, et al.
Journal of Hazardous Materials, 285, 311-318 (2015)
Determination of iodine in natural waters (sodium chloride as a reagent in the catalytic reduction of ceric ions).
Dubravcic M.
Analyst, 80(949), 295-300 (1955)
Analysis of salt penetration enhancement in meat tissue by mechanical treatment using a tumbling simulator.
Sharedeh D, et al.
Journal of Food Engineering, 166, 377-383 (2015)
Sodium chloride, a cost effective partial replacement of calcium ascorbate and ascorbic acid to inhibit surface browning on fresh-cut apple slices
Li, Y,et al.
LWT--Food Science and Technology, 64(1), 503-507 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service