Skip to Content
Merck
All Photos(1)

Key Documents

423599

Sigma-Aldrich

2-Ethyl-4-methylthiazole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NS
CAS Number:
Molecular Weight:
127.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.505 (lit.)

bp

161-162 °C (lit.)

density

1.026 g/mL at 25 °C (lit.)

SMILES string

CCc1nc(C)cs1

InChI

1S/C6H9NS/c1-3-6-7-5(2)4-8-6/h4H,3H2,1-2H3

InChI key

VGRVKVGGUPOCMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethyl-4-methylthiazole is a thiazole derivative. It is reported to be one of the aromatic volatile compound formed during the Maillard reaction between reaction L-ascorbic acid and L-cysteine. Its palladium-catalyzed direct coupling reaction with 3-bromochromen-4-one has been examined.

Application

2-Ethyl-4-methylthiazole may be used in the synthesis of the following:
  • 4-(2-ethyl-4-methylthiazol-5-yl)benzaldehyde
  • 2-(2-ethyl-4-methylthiazol-5-yl)benzonitrile
  • 4-(2-ethyl-4-methylthiazol-5-yl)-pyridine

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aroma compounds generated from thermal reaction of L-ascorbic acid with L-cysteine.
Yu AN and Zhang AD.
Food Chemistry, 121(4), 1060-1065 (2010)
Carbonates: eco-friendly solvents for palladium-catalysed direct arylation of heteroaromatics.
Dong JJ, et al.
Green Chemistry, 12(11), 2053-2063 (2010)
Pd-catalysed heteroarylations of 3-bromochromen-4-one via C-H bond activation of heteroarenes.
Belkessam F, et al.
Tetrahedron Letters, 54(36), 4888-4891 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service