Skip to Content
Merck
All Photos(1)

Key Documents

403121

Sigma-Aldrich

Poly(3-hydroxybutyric acid-co-3-hydroxyvaleric acid)

natural origin, PHV content 12 mol %

Synonym(s):

PHBV

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[COCH2CH(CH3)O]m[COCH2CH(C2H5)O]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Pricing and availability is not currently available.

description

12 mol% is approximately 14 wt%

optical activity

[α]20/D +11.8°, c = 0.1 in chloroform

composition

PHV content, 12 mol %

SMILES string

CC(O)CC(O)=O.CCC(O)CC(O)=O

InChI

1S/C5H10O3.C4H8O3/c1-2-4(6)3-5(7)8;1-3(5)2-4(6)7/h4,6H,2-3H2,1H3,(H,7,8);3,5H,2H2,1H3,(H,6,7)

InChI key

IUPHTVOTTBREAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(3-hydroxybutyric acid-co-3-hydroxyvaleric acid) is a biocompatible and biodegradable polymer which is produced naturally by a number of microorganisms.[1]

Application

PHBV based foam matrix may be used to produce tissue engineered bone.[1] It was used as a standard for calibration in another study which focused on the effect of fatty acids on PHA producing microorganism communities.[2]

Features and Benefits

Biodegradable polymer

Preparation Note

Produced via a controlled fermentation process using microoroganisms.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile fatty acids influence on the structure of microbial communities producing PHAs.
Ciesielski S and Przybylek G
Brazilian Journal of Microbiology, 45(2), 395-402 (2014)
Poly (3-hydroxybutyric acid-co-3-hydroxyvaleric acid) based tissue engineering matrices.
Kose G T, et al.
Journal of Materials Science. Materials in Medicine, 14(2), 121-126 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service