Skip to Content
Merck
All Photos(1)

Key Documents

400653

Sigma-Aldrich

Sodium niobate

−100 mesh, 99.9%

Synonym(s):

Niobium sodium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaNbO3
CAS Number:
Molecular Weight:
163.89
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.9%

form

powder

reaction suitability

reagent type: catalyst
core: niobium

particle size

−100 mesh

SMILES string

[Na+].[O-][Nb](=O)=O

InChI

1S/Na.Nb.3O/q+1;;;;-1

InChI key

UYLYBEXRJGPQSH-UHFFFAOYSA-N

Application

Reagent / Reactant for preparation of:
  • Transparent (Sr,Ba)Nb2O6 ceramics with optoelectric properties
  • Niobium sodium strontium oxides
  • Tungsten bronze for study of sodium/proton and sodium/deuteron cation exchange
  • Triple-layered Aurivillius phase photocatalysts
  • Niobium sodium selenide/sulfide
  • Bismuth niobium sodium oxides
  • Tin niobates

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service