Skip to Content
Merck
All Photos(1)

Key Documents

400149

Sigma-Aldrich

(1R,2R)-1,2-Dicyclohexyl-1,2-ethanediol

95%

Synonym(s):

(1R,2R)-1,2-Dicyclohexylethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H11CH(OH)-]2
CAS Number:
Molecular Weight:
226.36
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

optical activity

[α]20/D −2.2°, c = 0.8 in chloroform

mp

138-142 °C (lit.)

SMILES string

O[C@H](C1CCCCC1)[C@H](O)C2CCCCC2

InChI

1S/C14H26O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h11-16H,1-10H2/t13-,14-/m1/s1

InChI key

CKHLARJSOMXDKL-ZIAGYGMSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

C2 symmetric chiral diol with versatile applications as a chiral auxiliary, building block, and chiral ligand. Chiral auxiliary for preparing highly diastereomerically pure alkyl boronates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemische Berichte, 122, 1783-1783 (1989)
Burk, M.J. et al.
Tetrahedron Asymmetry, 2, 569-569 (1991)
Marshall, J.A. Xie, S.
The Journal of Organic Chemistry, 60, 7230-7230 (1995)
Stolle, A. et al.
Tetrahedron Letters, 35, 3521-3521 (1994)
Ho, O.C. et al.
Organometallics, 14, 2855-2855 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service