Skip to Content
Merck
All Photos(1)

Documents

391611

Sigma-Aldrich

7,8,9,10-Tetrahydrobenzo[a]pyren-7-ol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16O
CAS Number:
Molecular Weight:
272.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

142-144 °C (lit.)

SMILES string

OC1CCCc2c1cc3ccc4cccc5ccc2c3c45

InChI

1S/C20H16O/c21-18-6-2-5-15-16-10-9-13-4-1-3-12-7-8-14(11-17(15)18)20(16)19(12)13/h1,3-4,7-11,18,21H,2,5-6H2

InChI key

VKUQFYXBPGXTKI-UHFFFAOYSA-N

General description

7,8,9,10-Tetrahydrobenzo[a]pyren-7-ol (7-hydroxy-7,8,9,10-tetrahydrobenzo[a]pyrene), a polycyclic aromatic hydrocarbon (PAH), is a substituted benzopyrene. Its bromination reaction with N-bromosaccharin (NBSac) has been investigated. The sulfotransferase-assisted activation of 7-hydroxy-7,8,9,10-tetrahydrobenzo[a]pyrene to form electrophilic, mutagenic, or tumorigenic sulfuric acid ester metabolite, which binds covalently to DNA and leads to mutation, has been reported.

Application

7,8,9,10-Tetrahydrobenzo[a]pyren-7-ol may be used as a starting material in the synthesis of (±)-trans-7,8-dihydroxy-6-fluoro-7,8-dihydrobenzo[a]pyrene.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y J Surh et al.
Chemical research in toxicology, 8(5), 693-698 (1995-07-01)
Some hydroxymethyl-substituted polycyclic aromatic hydrocarbons have been shown to be converted to electrophilic, mutagenic, or tumorigenic sulfuric acid ester metabolites by cytosolic sulfotransferase activity in rodent liver. Likewise, certain types of aromatic compounds with a secondary alcoholic functional group at
V Heisig et al.
Science (New York, N.Y.), 223(4633), 289-291 (1984-01-20)
The laser excited fluorescence-line-narrowed spectrum of DNA modified with (+/-)-r-7,t-8-dihydroxy-t-9,10-epoxy-7,8,9,10-tetrahydrobenzo[a]pyrene (BPDE), the ultimate carcinogenic metabolite of benzo[a]pyrene (BP), has been obtained in a water-glycerol-ethanol glass at 4.2 K. The spectrum was well resolved and highly characteristic of the chromophore. Comparisons
Recent Progress in the Use of N-Halo Compounds in Organic Synthesis.
Veisi H, et al.
Organic preparations and procedures international, 43(6), 489-540 (2011)
Barbara Zajc
The Journal of organic chemistry, 64(6), 1902-1907 (2001-10-25)
Synthesis of (+/-)-trans-7,8-dihydroxy-6-fluoro-7,8-dihydrobenzo[a]pyrene, the metabolite from 6-fluoro-benzo[a]pyrene, is described. Position 6 of 7,8,9,10-tetrahydrobenzo[a]pyren-7-ol (1) was functionalized by bromination with N-bromosaccharin. Regioselectivity in the bromination is thought to derive from a substrate-reagent hydrogen bond. NMR evidence is offered to support this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service