Skip to Content
Merck
All Photos(1)

Key Documents

374741

Sigma-Aldrich

Pectic acid

Synonym(s):

Pectic acid from apple, Poly-D-galacturonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H8O6]n
CAS Number:
MDL number:
UNSPSC Code:
12352103

Application

For the removal of alkaline earth and heavy metal ions from wine. For x-ray diffraction analysis of pectic acid.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Wucherpfennig, et al.
Wein-Wiss., 30, 132-132 (1984)
Conformation and interactions of pectins. I. X-ray diffraction analyses of sodium pectate in neutral and acidified forms.
M D Walkinshaw et al.
Journal of molecular biology, 153(4), 1055-1073 (1981-12-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service