Skip to Content
Merck
All Photos(1)

Key Documents

373281

Sigma-Aldrich

Platinum-rhodium alloy

Rhodium 6 %

Synonym(s):

Pt41Rh5

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

thermocouple grade

form

wire

composition

Rhodium, 6%

diam.

0.51 mm

SMILES string

[Rh].[Pt]

InChI

1S/Pt.Rh

InChI key

PXXKQOPKNFECSZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

2.1 g = 50 cm

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service