Skip to Content
Merck
All Photos(1)

Documents

366420

Sigma-Aldrich

Tributylstannyl trifluoromethanesulfonate

90%

Synonym(s):

Tributyltin triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3Sn[(CH2)3CH3]3
CAS Number:
Molecular Weight:
439.12
Beilstein:
5818090
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

SMILES string

CCCC[Sn](CCCC)(CCCC)OS(=O)(=O)C(F)(F)F

InChI

1S/3C4H9.CHF3O3S.Sn/c3*1-3-4-2;2-1(3,4)8(5,6)7;/h3*1,3-4H2,2H3;(H,5,6,7);/q;;;;+1/p-1

InChI key

LIQOILBASIAIQC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service