Skip to Content
Merck
All Photos(1)

Key Documents

365955

Sigma-Aldrich

Niobium(III) bromide ethylene glycol dimethyl ether complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NbBr3O2C4H10
CAS Number:
Molecular Weight:
422.74
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

solid

mp

155 °C (dec.) (lit.)

SMILES string

Br[Nb](Br)Br.COCCOC

InChI

1S/C4H10O2.3BrH.Nb/c1-5-3-4-6-2;;;;/h3-4H2,1-2H3;3*1H;/q;;;;+3/p-3

InChI key

XDLURHXPYYXULL-UHFFFAOYSA-K

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service