Skip to Content
Merck
All Photos(1)

Documents

362360

Sigma-Aldrich

3-Methoxyacrylonitrile, mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH=CHCN
CAS Number:
Molecular Weight:
83.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.455 (lit.)

density

0.99 g/mL at 25 °C (lit.)

SMILES string

COC=CC#N

InChI

1S/C4H5NO/c1-6-4-2-3-5/h2,4H,1H3

InChI key

IPCRTSDORDQHRO-UHFFFAOYSA-N

Application

3-Methoxyacrylonitrile may be used in the synthesis of 5-amino-1-hydroxyethylpyrazole, an intermediate of cefoselis sulfate. It may be used in the synthesis of 3-aminoacrylonitrile, an important intermediate for the synthesis of biologically active substances.2

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 5-Amino-1-hydroxyethylpyrazole.
Zhon Y and Wu B.
Chinese Journal of Pharmaceuticals / Chung-kuo i yao kung yeh tsa chih, 34(9), 431-432 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service