Skip to Content
Merck
All Photos(1)

Key Documents

344567

Sigma-Aldrich

2-Ethoxyanisole

98%

Synonym(s):

1-Ethoxy-2-methoxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4OCH3
CAS Number:
Molecular Weight:
152.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.524 (lit.)

bp

217-218 °C (lit.)

density

1.044 g/mL at 25 °C (lit.)

SMILES string

CCOc1ccccc1OC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service