317322
(S)-(−)-1-Phenyl-1-butanol
97%
Synonym(s):
(S)-(−)-α-Propylbenzyl alcohol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH2CH2CH(C6H5)OH
CAS Number:
Molecular Weight:
150.22
Beilstein:
2043698
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
Recommended Products
Assay
97%
optical activity
[α]21/D −48.6°, c = 5 in chloroform
optical purity
ee: 99% (GLC)
bp
120 °C/0.05 mmHg (lit.)
mp
45-47 °C (lit.)
SMILES string
CCC[C@H](O)c1ccccc1
InChI
1S/C10H14O/c1-2-6-10(11)9-7-4-3-5-8-9/h3-5,7-8,10-11H,2,6H2,1H3/t10-/m0/s1
InChI key
HQRWWHIETAKIMO-JTQLQIEISA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
217.4 °F - closed cup
Flash Point(C)
103 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service