30763
Boc-α-(diphenylmethyl)-DL-Pro-OH
technical, ≥90% (HPLC)
Synonym(s):
1-Boc-2-(diphenylmethyl)-2-pyrrolidinecarboxylic acid, Boc-α-(benzhydryl)-DL-proline, Boc-α-(diphenylmethyl)-DL-proline
About This Item
Recommended Products
grade
technical
Assay
≥90% (HPLC)
form
powder
reaction suitability
reaction type: Boc solid-phase peptide synthesis
application(s)
peptide synthesis
SMILES string
CC(C)(C)OC(=O)N1CCCC1(C(c2ccccc2)c3ccccc3)C(O)=O
InChI
1S/C23H27NO4/c1-22(2,3)28-21(27)24-16-10-15-23(24,20(25)26)19(17-11-6-4-7-12-17)18-13-8-5-9-14-18/h4-9,11-14,19H,10,15-16H2,1-3H3,(H,25,26)
InChI key
RUBYLACKGJPIPO-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service