Skip to Content
Merck
All Photos(1)

Key Documents

281530

Sigma-Aldrich

Montmorillonite KSF

Synonym(s):

K-Catalyst

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352106

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

  1. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  2. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  3. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  4. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  5. Is it possible for Montmorillonite KSF, Product 281530, to contain trace amounts of metallic components?

    Yes. According to our supplier, it is possible for trace metals to be present. The most likely metallic component present is iron; however, barium, chromium, manganese, and nickel can also be present. However, Sigma-Aldrich does not perform quantitative metals testing on this material.

  6. How can Montmorillonite KSF, Product 281530, be used?

    K-type montmorillonites are acidic catalysts that can be used in some synthesis reactions in place of Bronsted acids (like H2SO4, HCl, H3PO4, HClO4, etc), or Lewis Acids (like BF3, BCl3, AlCl3, SnCl4, etc.). Although the pH is not tested by Sigma-Aldrich, it is reported to be about 1.5 for a 10% aqueous suspension of the clay catalyst. Montmorillonite KSF is easier to handle and less corrosive than the traditional aforementioned acid catalysts.

  7. My question is not addressed here, how can I contact Technical Service for assistance?

    Ask a Scientist here.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service