264709
Isobutyl acetate
95%, natural
Synonym(s):
Acetic acid isobutyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3COOCH2CH(CH3)2
CAS Number:
Molecular Weight:
116.16
Beilstein:
1741909
EC Number:
MDL number:
UNSPSC Code:
12352103
grade
natural
vapor density
>4 (vs air)
vapor pressure
15 mmHg ( 20 °C)
Assay
95%
autoignition temp.
793 °F
expl. lim.
10.5 %
refractive index
n20/D 1.39 (lit.)
bp
115-117 °C (lit.)
mp
−99 °C (lit.)
density
0.867 g/mL at 25 °C (lit.)
SMILES string
CC(C)COC(C)=O
InChI
1S/C6H12O2/c1-5(2)4-8-6(3)7/h5H,4H2,1-3H3
InChI key
GJRQTCIYDGXPES-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service