Skip to Content
Merck
All Photos(1)

Key Documents

245690

Sigma-Aldrich

2,4,6-Tribromoresorcinol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br3C6H-1,3-(OH)2
CAS Number:
Molecular Weight:
346.80
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

114-116 °C (lit.)

functional group

bromo

SMILES string

Oc1c(Br)cc(Br)c(O)c1Br

InChI

1S/C6H3Br3O2/c7-2-1-3(8)6(11)4(9)5(2)10/h1,10-11H

InChI key

YADZSMVDNYXOOB-UHFFFAOYSA-N

General description

2,4,6-Tribromoresorcinol was quantified by using five-point calibration curves with data points.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian Weisz et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 31(6), 979-984 (2014-03-22)
Specifications in the US Code of Federal Regulations (CFR) for the colour additives D&C Red No. 27 (R27) and D&C Red No. 28 (R28) limit the levels of the synthetic by-products 2,3,4,5-tetrachloro-6-(3´,5´-dibromo-2´,4´-dihydroxybenzoyl)benzoic acid (SBBA) and "brominated resorcinol" and of the
Nebebech Belai et al.
Journal of AOAC International, 102(3), 936-941 (2018-11-20)
Background: Certain impurities in the color additives drug and cosmetic (D&C) Red No. 21 (R21), D&C Red No. 22 (R22), and their lakes are limited to levels specified in the Code of Federal Regulations (CFR) and are quantified by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service