Skip to Content
Merck
All Photos(1)

Documents

244317

Sigma-Aldrich

Saccharin sodium salt hydrate

≥99%

Synonym(s):

2,3-Dihydro-3-oxobenzisosulfonazole sodium salt, 2-Sulfobenzoic acid imide sodium salt, o-Sulfobenzimide sodium salt, Saccharin soluble

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4NNaO3S · xH2O
CAS Number:
Molecular Weight:
205.17 (anhydrous basis)
Beilstein:
3599229
MDL number:
UNSPSC Code:
12352103

Assay

≥99%

SMILES string

[Na+].[H]O[H].[O-]C1=NS(=O)(=O)c2ccccc12

InChI

1S/C7H5NO3S.Na.H2O/c9-7-5-3-1-2-4-6(5)12(10,11)8-7;;/h1-4H,(H,8,9);;1H2/q;+1;/p-1

InChI key

ILJOYZVVZZFIKA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service