Skip to Content
Merck
All Photos(1)

Key Documents

241164

Sigma-Aldrich

Tetraethylene glycol dimethyl ether

≥99%

Synonym(s):

2,5,8,11,14-Pentaoxapentadecane, Bis[2-(2-methoxyethoxy)ethyl] ether, Dimethoxytetraethylene glycol, Dimethyltetraglycol, Tetraglyme

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)4CH3
CAS Number:
Molecular Weight:
222.28
Beilstein:
1760005
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor density

7.7 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

510 °F

reaction suitability

reagent type: cross-linking reagent
reaction type: click chemistry

refractive index

n20/D 1.432 (lit.)

bp

275-276 °C (lit.)

mp

−30 °C (lit.)

density

1.009 g/mL at 25 °C (lit.)

polymer architecture

shape: linear
functionality: homobifunctional

SMILES string

COCCOCCOCCOCCOC

InChI

1S/C10H22O5/c1-11-3-5-13-7-9-15-10-8-14-6-4-12-2/h3-10H2,1-2H3

InChI key

ZUHZGEOKBKGPSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lan Cao et al.
Journal of biomedical materials research. Part A, 81(1), 12-23 (2006-11-17)
In this study, a glow discharge plasma deposition system previously used for treating flat substrates was successfully modified and optimized to produce a PEO-like coating on the inner surface of 1-3 mm ID polyethylene tubing by deposition of tetra ethylene
Lan Cao et al.
Journal of biomedical materials research. Part A, 79(4), 788-803 (2006-08-03)
Previous studies from our lab have shown that fibrinogen adsorption (Gamma(Fg)) must be reduced below 10 ng/cm(2) to significantly reduce platelet adhesion, and that radio frequency glow discharge (RFGD) treatment of polymeric films in the presence of tetraethylene glycol dimethyl
Malinda Salim et al.
Lab on a chip, 7(4), 523-525 (2007-03-29)
This Technical Note presents the direct surface modification of a glass/PTFE hybrid microfluidic chip, via radio frequency glow discharge plasma polymerisation of tetraethlylene glycol dimethylether (tetraglyme), to produce hydrophilic, non-fouling, PEO-like surfaces. We use several techniques including X-ray photoelectron spectroscopy
Hun-Gi Jung et al.
Nature chemistry, 4(7), 579-585 (2012-06-22)
Although dominating the consumer electronics markets as the power source of choice for popular portable devices, the common lithium battery is not yet suited for use in sustainable electrified road transport. The development of advanced, higher-energy lithium batteries is essential
Claire R Hurley et al.
Langmuir : the ACS journal of surfaces and colloids, 26(12), 10203-10209 (2010-03-25)
Plasma-polymerized tetraglyme films (PP4G) have been modified by exposure to ultraviolet (UV) light from a frequency-doubled argon ion laser (244 nm) and characterized using X-ray photoelectron spectroscopy (XPS) and atomic force microscopy (AFM). XPS data indicated that the ether component

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service