Skip to Content
Merck
All Photos(1)

Key Documents

234656

Sigma-Aldrich

Fast Garnet GBC sulfate salt

purified, Dye content 75 %

Synonym(s):

2-Methyl-4-([2-methylphenyl]azo)benzenediazonium salt, 4-Amino-2′,3-dimethylazobenzene diazotated, Azoic Diazo No. 4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N4O4S
CAS Number:
Molecular Weight:
334.35
Colour Index Number:
37210
EC Number:
MDL number:
UNSPSC Code:
12171500

composition

Dye content, 75%

λmax

360 nm

SMILES string

OS([O-])(=O)=O.Cc1ccccc1N=Nc2ccc([N+]#N)c(C)c2

InChI

1S/C14H13N4.H2O4S/c1-10-5-3-4-6-14(10)18-17-12-7-8-13(16-15)11(2)9-12;1-5(2,3)4/h3-9H,1-2H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

PPTXFSQSISUDAU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service