Skip to Content
Merck
All Photos(1)

Key Documents

228311

Sigma-Aldrich

Potassium cyclohexanebutyrate

Synonym(s):

Cyclohexanebutyric acid potassium salt, Potassium 4-cyclohexylbutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11(CH2)3CO2K
CAS Number:
Molecular Weight:
208.34
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

95.0-105.0% (perchloric acid titration)

form

crystals and lumps

contains

≤5% cyclohexanebutyric acid for enhanced solubility

SMILES string

[K+].[O-]C(=O)CCCC1CCCCC1

InChI

1S/C10H18O2.K/c11-10(12)8-4-7-9-5-2-1-3-6-9;/h9H,1-8H2,(H,11,12);/q;+1/p-1

InChI key

LBYNWSSEILNGLN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Potassium standard salt for organic systems. Lot specific assay on label.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H J Ougham et al.
Journal of bacteriology, 150(3), 1172-1182 (1982-06-01)
A strain of Arthrobacter was isolated by enrichment culture with cyclohexaneacetate as the sole source of carbon and grew with a doubling time of 4.2 h. In addition to growing with cyclohexaneacetate, the organism also grew with cyclohexanebutyrate at concentrations
Dean M Quesnel et al.
Chemosphere, 84(4), 504-511 (2011-04-05)
Naphthenic acids (NAs) are a major contributor to toxicity in tailings waste generated from bitumen production in the Athabasca Oil Sands region. While investigations have shown that bacteria can biodegrade NAs and reduce tailings toxicity, the potential of algae to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service