Skip to Content
Merck
All Photos(1)

Documents

19079

Sigma-Aldrich

3,4-(Methylenedioxy)mandelic acid

≥98.0%

Synonym(s):

α-Hydroxy-1,3-benzodioxole-5-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8O5
CAS Number:
Molecular Weight:
196.16
Beilstein:
209546
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0%

form

powder

mp

158-162 °C

SMILES string

OC(C(O)=O)c1ccc2OCOc2c1

InChI

1S/C9H8O5/c10-8(9(11)12)5-1-2-6-7(3-5)14-4-13-6/h1-3,8,10H,4H2,(H,11,12)

InChI key

CLUJFRCEPFNVHW-UHFFFAOYSA-N

Application

3,4-(Methylenedioxy)mandelic acid was used as starting reagent in the synthesis of (-)cephalotaxine, (+)harringtonine and (-)drupacine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hajer Abdelkafi et al.
Natural product reports, 29(8), 845-869 (2012-06-21)
The Cephalotaxus genus belongs to the Cephalotaxaceae family of conifers. Over the past decades it has proved to be a fruitful source of interesting natural products, especially alkaloids (cephalotaxine esters) and terpenoids (abietanes, troponoids), which often display medicinal properties, especially
Anna Constance Vind et al.
Molecular cell, 78(4), 700-713 (2020-04-15)
Impairment of ribosome function activates the MAPKKK ZAK, leading to activation of mitogen-activated protein (MAP) kinases p38 and JNK and inflammatory signaling. The mechanistic basis for activation of this ribotoxic stress response (RSR) remains completely obscure. We show that the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service