Skip to Content
Merck
All Photos(2)

Documents

17380

Sigma-Aldrich

Fluoro-N,N,N′,N′-bis(tetramethylene)formamidinium hexafluorophosphate

≥99.0% (T), for peptide synthesis

Synonym(s):

BTFFH, Fluoro-dipyrrolidinocarbenium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16F7N2P
CAS Number:
Molecular Weight:
316.20
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

product name

Fluoro-N,N,N′,N′-bis(tetramethylene)formamidinium hexafluorophosphate, ≥99.0% (T)

Assay

≥99.0% (T)

form

powder

reaction suitability

reaction type: Coupling Reactions

mp

154-160 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

F[P-](F)(F)(F)(F)F.F\C(N1CCCC1)=[N+]2/CCCC2

InChI

1S/C9H16FN2.F6P/c10-9(11-5-1-2-6-11)12-7-3-4-8-12;1-7(2,3,4,5)6/h1-8H2;/q+1;-1

InChI key

MNJUGQKOHJQOCK-UHFFFAOYSA-N

Application

Reagent for synthesis of immonium-type coupling reagents

Other Notes

Convenient peptide coupling reagent

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. El-Faham
Chemistry Letters (Jpn), 671-671 (1998)
L.A. Carpino et al.
Journal of the American Chemical Society, 117, 5401-5401 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service