Skip to Content
Merck
All Photos(1)

Key Documents

15477

Sigma-Aldrich

Boc-Cha-OH hydrate

≥99.0% (TLC), ~1 mol/mol water

Synonym(s):

Boc-3-cyclohexyl-L-alanine hydrate, Boc-hexahydro-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H25NO4 · xH2O
CAS Number:
Molecular Weight:
271.35 (anhydrous basis)
Beilstein:
3060603
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99.0% (TLC)

optical activity

[α]20/D −15.5±1°, c = 2% in glacial acetic acid

impurities

~1 mol/mol water

application(s)

peptide synthesis

SMILES string

O.CC(C)(C)OC(=O)N[C@@H](CC1CCCCC1)C(O)=O

InChI

1S/C14H25NO4.H2O/c1-14(2,3)19-13(18)15-11(12(16)17)9-10-7-5-4-6-8-10;/h10-11H,4-9H2,1-3H3,(H,15,18)(H,16,17);1H2/t11-;/m0./s1

InChI key

KGIDHDHQFLWAMT-MERQFXBCSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service