Skip to Content
Merck
All Photos(2)

Documents

148679

Sigma-Aldrich

5-Vinyl-2-norbornene, mixture of endo and exo

95%, contains 80-150 ppm BHT as inhibitor

Synonym(s):

2-Vinyl-5-norbornene, 2-Vinylbicyclo[2.2.1]hept-5-ene, 2-Vinylnorbornene, 5-Ethenylbicyclo[2.2.1]hept-2-ene, 5-Vinylbicyclo[2.2.1]hept-2-ene, 5-Vinylnorbornene, 6-Vinylbicyclo[2.2.1]hept-2-ene, Vinylnorbornene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12
CAS Number:
Molecular Weight:
120.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

vapor pressure

6 mmHg ( 20 °C)

Assay

95%

form

liquid

contains

80-150 ppm BHT as inhibitor

refractive index

n20/D 1.481 (lit.)

bp

141 °C (lit.)

mp

−80 °C (lit.)

density

0.841 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CC1C[C@H]2C[C@@H]1C=C2

InChI

1S/C9H12/c1-2-8-5-7-3-4-9(8)6-7/h2-4,7-9H,1,5-6H2/t7-,8?,9-/m0/s1

InChI key

INYHZQLKOKTDAI-SMOXQLQSSA-N

Related Categories

Application

5-Vinyl-2-norbornene was used in the preparation of 5-ethylidene-2-norbornene (ENB).

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hoon Sik Kim et al.
The Journal of organic chemistry, 71(3), 911-914 (2006-01-28)
The isomerization reaction of 5-vinyl-2-norbornene (VNB) to 5-ethylidene-2-norbornene (ENB) has been performed using a catalytic system consisting of an alkali metal hydride and an amine. Among various amines tested, only aliphatic 1,2-diamines exhibited the activity for the isomerization. The isomerization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service