Skip to Content
Merck
All Photos(1)

Key Documents

130761

Sigma-Aldrich

(3′-Nitropropiophenone)

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4COC2H5
CAS Number:
Molecular Weight:
179.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

98-101 °C (lit.)

functional group

ketone
nitro

SMILES string

CCC(=O)c1cccc(c1)[N+]([O-])=O

InChI

1S/C9H9NO3/c1-2-9(11)7-4-3-5-8(6-7)10(12)13/h3-6H,2H2,1H3

InChI key

VSPOTMOYDHRALZ-UHFFFAOYSA-N

Application

3′-Nitropropiophenone was used to study the FTIR and FT Raman spectra in the regions 3500-100 cm(-1).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Udhayakala et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 75(2), 567-573 (2009-12-25)
The FTIR and FT Raman spectra of 3'-chloropropiophenone and 3'-nitropropiophenone have been recorded in the regions 4000-400 and 3500-100 cm(-1) respectively. The optimized geometry, frequency and intensity of the vibrational bands of 3'-chloropropiophenone and 3'-nitropropiophenone were obtained by ab initio

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service