Skip to Content
Merck
All Photos(1)

Key Documents

110396

Supelco

Chromotrope 2B

Synonym(s):

2-(4-Nitrophenylazo)chromotropic acid disodium salt, Acid Red 176

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H9N3Na2O10S2
CAS Number:
Molecular Weight:
513.37
Colour Index Number:
16575
Beilstein:
4109733
EC Number:
MDL number:
UNSPSC Code:
12352200

SMILES string

[Na+].[Na+].Oc1cc(cc2cc(c(\N=N\c3ccc(cc3)[N+]([O-])=O)c(O)c12)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C16H11N3O10S2.2Na/c20-12-7-11(30(24,25)26)5-8-6-13(31(27,28)29)15(16(21)14(8)12)18-17-9-1-3-10(4-2-9)19(22)23;;/h1-7,20-21H,(H,24,25,26)(H,27,28,29);;/q;2*+1/p-2/b18-17+;;

InChI key

AXUUHWJXDWBCSG-QIKYXUGXSA-L

Looking for similar products? Visit Product Comparison Guide

Application

For the determination of boric acid, borates, germanium, cobalt, nickel, copper and rare earth metals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service