Skip to Content
Merck
All Photos(1)

Key Documents

108936

Sigma-Aldrich

2-Methylbenzoxazole

99%

Synonym(s):

2-Methyl-1,3-benzoxazole, 2-Methyl-4,5-benzoxazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NO
CAS Number:
Molecular Weight:
133.15
Beilstein:
112297
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.550 (lit.)

bp

178 °C (lit.)

mp

5-10 °C (lit.)

density

1.121 g/mL at 25 °C (lit.)

SMILES string

Cc1nc2ccccc2o1

InChI

1S/C8H7NO/c1-6-9-7-4-2-3-5-8(7)10-6/h2-5H,1H3

InChI key

DQSHFKPKFISSNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

2-Methylbenzoxazole condenses with aromatic aldehyde having acidic protons during the synthesis of HIV-reverse transcriptase inhibitor L-696,299. 2-Methylbenzoxazole has been used in synthesis of bis-styryl dyes.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Styryl dyes-synthesis and applications during the last 15 years.
Deligeorgiev T, et al.
Coloration Technology, 126(2), 55-80 (2010)
Condensation of 2-methylbenzoxazole with aromatic aldehydes bearing acidic protons. A convenient coupling in the synthesis of the HIV-reverse transcriptase inhibitor L-696,229.
Houpis IN, et al.
The Journal of Organic Chemistry, 58(11), 3176-3178 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service