ADVH99BD5B63
AChemBlock
N1,N1,N2,N2-tetramethylpropane-1,2-diamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
130.23
Iupac Name
N~1~,N~1~,N~2~,N~2~-tetramethyl-1,2-propanediamine
Inchi Code
1S/C7H18N2/c1-7(9(4)5)6-8(2)3/h7H,6H2,1-5H3
InChI key
JUXXCHAGQCBNTI-UHFFFAOYSA-N
Purity
0.97
Country of Origin
CN
Product Link
https://www.achemblock.com/x208462-n1-n1-n2-n2-tetramethylpropane-1-2-diamine.html
Synonym
N1,N1,N2,N2-tetramethylpropane-1,2-diamine
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.achemblock.com/x208462-n1-n1-n2-n2-tetramethylpropane-1-2-diamine.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.