Skip to Content
Merck
All Photos(1)

Key Documents

ADVH987334AD

AChemBlock

2,8-dioxaspiro[4.5]decane-1,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
170.16
MDL number:

Iupac Name

2,8-dioxaspiro[4.5]decane-1,3-dione

Inchi Code

1S/C8H10O4/c9-6-5-8(7(10)12-6)1-3-11-4-2-8/h1-5H2

InChI key

BPLDSHSHEFQZPP-UHFFFAOYSA-N

storage temp.

Store at Room Tem.

Purity

0.97

Country of Origin

CN

Physical Form

off-white solid

Product Link

https://www.achemblock.com/q59469-2-8-dioxaspiro-4-5-decane-1-3-dione.html

Synonym

2,8-dioxaspiro[4.5]decane-1,3-dione

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.achemblock.com/q59469-2-8-dioxaspiro-4-5-decane-1-3-dione.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library