Skip to Content
Merck
All Photos(1)

Key Documents

AABH97CE87A5

AA BLOCKS, INC.

(3-nitrophenyl)(pyrrolidin-1-yl)acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
231.25
MDL number:

Iupac Name

(3-nitrophenyl)(1-pyrrolidinyl)acetonitrile

Inchi Code

1S/C12H13N3O2/c13-9-12(14-6-1-2-7-14)10-4-3-5-11(8-10)15(16)17/h3-5,8,12H,1-2,6-7H2

InChI key

CTMPSAKTWCHPSJ-UHFFFAOYSA-N

storage temp.

2~8 C

Purity

95%

Country of Origin

US

Product Link

https://www.aablocks.com/prod/687614-64-8

Synonym

(3-Nitrophenyl)(pyrrolidin-1-yl)acetonitrile; .

MSDS

https://www.aablocks.com/downloadSds?cas=687614-64-8


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library