Merck
  • Bio-Compatible Ca-BDC/Polymer Monolithic Composites Templated from Bio-Active Ca-BDC Co-Stabilized CO2-in-Water High Internal Phase Emulsions.
Bio-Compatible Ca-BDC/Polymer Monolithic Composites Templated from Bio-Active Ca-BDC Co-Stabilized CO2-in-Water High Internal Phase Emulsions.
Polymers (2020-04-23)
Xule Yang, Youwei Hao, Liqin Cao
ABSTRACT
Because of the nontoxic solvents contained in CO2-in-water emulsions, porous polymer composites templated from these emulsions are conducive for bio-applications. Herein, bio-active rod-like calcium-organic framworks (Ca-BDC MOFs, BDC= 1,4-benzenedicarboxylate anion) particles co-stabilized CO2-in-water high internal phase emulsion (C/W HIPE) in the presence of polyvinyl alcohol (PVA) is first presented. After curing of the continuous phase, followed by releasing CO2, integral 3D macro-porous Ca-BDC monolith and Ca-BDC/Poly(2-hydroxyethyl methacrylate-co-acrylamide) HIPEs monolithic composites [Ca-BDC/P(AM-co-HEMA)HIPEs] with open-cell macro-porous structures were successfully prepared. The pore structure of these porous composite can be tuned by means of tailoring the Ca-BDC dosage, carbon dioxide pressure, and continuous phase volume fractions in corresponding C/W HIPEs. Results of bio-compatibility tests show that these Ca-BDC/P(AM-co-HEMA)HIPEs monoliths have non-cytotoxicity on HepG2 cells; also, the E. coli can grow either on the surfaces or inside these monoliths. Furthermore, immobilization of β-amylase on these porous composite presents that β-amylase can be well-anchored into the porous polymer composites, its catalytic activity can be maintained even after 10 cycles. This work combined bio-active MOFs Ca-BDC, bio-compatible open-cell macroporous polymer PAM-co-HEMA and green C/W HIPEs to present a novel and facile way to prepare interconnected macro-porous MOFs/polymer composites. Compared with the existing other well-known materials such as hydrogels, these porous composites possess well-defined tunable pore structures and superior bio-activity, thereby have promising applications in bio-tissue engineering, food, and pharmaceutical.
Product Description
Supelco
Vials, screw top with solid green Thermoset cap with PTFE liner, preassembled, pkg of 100, volume 2 mL, amber glass vial (standard opening - 4.6 mm), O.D. × H × I.D. 12 mm × 32 mm × 4.6 mm, thread for 8-425